Assay Details
Target Gene Details
Entrez Gene ID: | 68505 |
Gene Name: | vacuolar protein sorting 51 homolog (S. cerevisiae) |
Gene Aliases: |
1110014N23Rik, 3110057M17Rik, AI837850, Ffr |
Location: |
Chr.19:6067842-6077187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps51 | NM_001081041.1 | NP_001074510.1 | ||
| AK137155.1 | BAE23255.1 | |||
| AK146211.1 | BAE26981.1 | |||
| AK180223.1 | ||||
| AK195187.1 | ||||
| AK216884.1 |

Genomic Map