Assay Details
Target Gene Details
Entrez Gene ID: | 110355 |
Gene Name: | adrenergic receptor kinase, beta 1 |
Gene Aliases: |
Adrbk-1, Bark-1, GRK2, beta ARK, betaARK1 |
Location: |
Chr.19:4285999-4306222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrbk1 | NM_001290818.1 | NP_001277747.1 | ||
| NM_130863.2 | NP_570933.1 | |||
| AF333028.1 | AAK21896.1 | |||
| AK153543.1 | BAE32080.1 | |||
| AK155691.1 | BAE33390.1 | |||
| BC003196.1 | AAH03196.1 | |||
| BC053922.1 | AAH53922.1 |
More Information
Set Membership: |
|

Genomic Map