Assay Details
Target Gene Details
Entrez Gene ID: | 66514 |
Gene Name: | asparaginase like 1 |
Gene Aliases: |
2410004D18Rik, ALP, ALP1, AU040643, AW060375 |
Location: |
Chr.19:9111719-9135566 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asrgl1 | NM_025610.3 | NP_079886.2 | ||
| AK006147.1 | BAB24431.1 | |||
| AK010381.1 | BAC25294.1 | |||
| AK030209.1 | BAC26844.1 | |||
| BC016106.1 | AAH16106.1 |
More Information
Set Membership: |
|

Genomic Map