Assay Details
Target Gene Details
Entrez Gene ID: | 546726 |
Gene Name: | cytochrome P450, family 26, subfamily c, polypeptide 1 |
Gene Aliases: |
EG546726 |
Location: |
Chr.19:37685680-37693307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp26c1 | NM_001105201.1 | NP_001098671.1 | ||
| BC151098.1 | ||||
| BC151106.1 |
More Information
Set Membership: |
|

Genomic Map