Assay Details
Target Gene Details
Entrez Gene ID: | 14544 |
Gene Name: | guanine deaminase |
Gene Aliases: |
AU015411, AW047581 |
Location: |
Chr.19:21391307-21472661 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gda | NM_010266.2 | 9 | 1045 | NP_034396.1 |
| AF174583.1 | 9 | 971 | AAD50297.1 | |
| AF369880.1 | 9 | 930 | AAM18369.1 | |
| AF369881.1 | 9 | 930 | AAM18370.1 | |
| AF369882.1 | 9 | 930 | AAM18371.1 | |
| AF369883.1 | 9 | 930 | AAM18372.1 | |
| AF369884.1 | 9 | 930 | AAM18373.1 | |
| AK041056.1 | 9 | 984 | BAC30802.1 | |
| AK079419.1 | 9 | 853 | ||
| AK079459.1 | 9 | 985 | ||
| AK157014.1 | 9 | 1007 | BAE33931.1 | |
| AK173138.1 | 9 | 974 | BAD32416.1 | |
| BC076583.1 | 9 | 958 | AAH76583.1 |
More Information
Set Membership: |
|

Genomic Map