Assay Details
Target Gene Details
Entrez Gene ID: | 14603 |
Gene Name: | gastric intrinsic factor |
Gene Aliases: |
AV073125 |
Location: |
Chr.19:11747543-11763447 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gif | NM_008118.3 | NP_032144.2 | ||
| AK078933.1 | BAC37468.1 | |||
| BC118519.1 | ||||
| L24191.1 | AAA37882.1 |

Genomic Map