Assay Details
Target Gene Details
Entrez Gene ID: | 71146 |
Gene Name: | golgi autoantigen, golgin subfamily a, 7B |
Gene Aliases: |
4933417O08Rik, AI839934 |
Location: |
Chr.19:42247578-42270348 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golga7b | NM_001141983.1 | NP_001135455.1 | ||
| AK016850.1 | BAB30463.1 |
More Information
Set Membership: |
|

Genomic Map