Assay Details
Target Gene Details
Entrez Gene ID: | 15958 |
Gene Name: | interferon-induced protein with tetratricopeptide repeats 2 |
Gene Aliases: |
AV302338, Ifi54, P54 |
Location: |
Chr.19:34550549-34576534 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifit2 | NM_008332.3 | NP_032358.1 | ||
| AK038027.1 | BAC29924.1 | |||
| AK042366.1 | BAC31239.1 | |||
| AK150206.1 | BAE29377.1 | |||
| AK150253.1 | BAE29415.1 | |||
| BC050835.1 | AAH50835.1 | |||
| CJ044173.1 | ||||
| U43085.1 | AAC52626.1 |

Genomic Map