Assay Details
Target Gene Details
Entrez Gene ID: | 14156 |
Gene Name: | flap structure specific endonuclease 1 |
Gene Aliases: |
AW538437, FEN-1 |
Location: |
Chr.19:10199132-10204187 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fen1 |

Genomic Map