Assay Details
Target Gene Details
Entrez Gene ID: | 23789 |
Gene Name: | coronin, actin binding protein 1B |
Gene Aliases: |
- |
Location: |
Chr.19:4148612-4154035 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coro1b |

Genomic Map