Assay Details
Target Gene Details
Entrez Gene ID: | 67894 |
Gene Name: | family with sequence similarity 45, member A |
Gene Aliases: |
1810055E12Rik |
Location: |
Chr.19:60811441-60836235 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam45a | NM_001167829.1 | NP_001161301.1 | ||
| NM_026437.3 | NP_080713.2 | |||
| AK004643.1 | ||||
| AK007867.1 | BAB25318.1 | |||
| AK145769.1 | BAE26640.1 | |||
| AK168474.1 | BAE40364.1 | |||
| AK171767.1 | 3 | 3069 | ||
| AK202624.1 | ||||
| BC013514.1 | AAH13514.1 | |||
| BY122265.1 |

Genomic Map