Assay Details
Target Gene Details
Entrez Gene ID: | 23789 |
Gene Name: | coronin, actin binding protein 1B |
Gene Aliases: |
- |
Location: |
Chr.19:4148612-4154035 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coro1b | NM_011778.2 | NP_035908.1 | ||
| AF143956.1 | AAD32704.1 | |||
| AK149639.1 | BAE29000.1 | |||
| AK159329.1 | ||||
| AK181268.1 | ||||
| AK209750.1 | ||||
| AK213023.1 | ||||
| BC131944.1 | ||||
| BC132617.1 |
More Information
Set Membership: |
|

Genomic Map