Assay Details
Target Gene Details
Entrez Gene ID: | 11632 |
Gene Name: | aryl-hydrocarbon receptor-interacting protein |
Gene Aliases: |
AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2 |
Location: |
Chr.19:4113756-4127236 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aip | NM_001276284.1 | 1 | 140 | NP_001263213.1 |
| NM_016666.3 | 2 | 239 | NP_057875.1 | |
| AK011609.1 | ||||
| AK146390.1 | 1 | 142 | BAE27133.1 | |
| BC075614.1 | AAH75614.1 | |||
| CX222057.1 | 1 | 109 | ||
| U85489.1 | 2 | 208 | AAB59009.1 |
More Information
Set Membership: |
|

Genomic Map