Assay Details
Target Gene Details
Entrez Gene ID: | 29866 |
Gene Name: | calcium binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.19:4081469-4087340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabp2 | NM_001160252.1 | NP_001153724.1 | ||
| NM_001160253.1 | NP_001153725.1 | |||
| NM_013878.2 | NP_038906.2 | |||
| AF169156.1 | 1 | 34 | AAF25790.1 | |
| AF169157.1 | 1 | 34 | AAF25791.1 | |
| AK007488.1 | ||||
| BC107029.2 | AAI07030.1 |

Genomic Map