Assay Details
Target Gene Details
Entrez Gene ID: | 73660 |
Gene Name: | calcium binding protein 4 |
Gene Aliases: |
2410038D05Rik |
Location: |
Chr.19:4135423-4144080 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabp4 | NM_144532.2 | 4 | 634 | NP_653115.1 |
| AK010631.1 | 1 | 176 | BAC25309.1 | |
| AK171863.1 | 7 | 2576 | ||
| AY039218.1 | 4 | 629 | AAK83463.1 | |
| BC049263.1 | 4 | 634 | AAH49263.1 |

Genomic Map