Assay Details
Target Gene Details
Entrez Gene ID: | 12868 |
Gene Name: | cytochrome c oxidase subunit VIIIa |
Gene Aliases: |
COX8L |
Location: |
Chr.19:7215158-7217616 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox8a | NM_007750.2 | 1 | 68 | NP_031776.1 |
| AK002218.1 | BAB21942.1 | |||
| AK002524.1 | BAB22161.1 | |||
| AK008594.1 | BAB25767.1 | |||
| BC120708.1 | ||||
| BC120710.1 | ||||
| U37721.1 | AAB38754.1 |
More Information
Set Membership: |
|

Genomic Map