Assay Details
Target Gene Details
Entrez Gene ID: | 72082 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 55 |
Gene Aliases: |
2010318C06Rik |
Location: |
Chr.19:39006938-39042687 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c55 | NM_028089.3 | NP_082365.1 | ||
| AK008580.1 | BAB25759.1 | |||
| AY206875.1 | AAO52738.1 | |||
| BC010824.1 | AAH10824.1 |
More Information
Set Membership: |
|

Genomic Map