Assay Details
Target Gene Details
Entrez Gene ID: | 68431 |
Gene Name: | F-box and leucine-rich repeat protein 15 |
Gene Aliases: |
0710008C12Rik, Fbxo37 |
Location: |
Chr.19:46328059-46330446 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl15 | NM_133694.2 | 2 | 383 | NP_598455.2 |
| AI594908.1 | ||||
| AK003032.1 | 2 | 107 | ||
| AK145009.1 | 2 | 341 | ||
| AK172339.1 | 3 | 280 | BAE42956.1 | |
| BC016499.1 | 2 | 358 | AAH16499.1 | |
| CK791452.1 | 2 | 69 |

Genomic Map