Assay Details
Target Gene Details
Entrez Gene ID: | 26358 |
Gene Name: | aldehyde dehydrogenase family 1, subfamily A7 |
Gene Aliases: |
AI987940, Ahd2-like, Aldh-pb, Aldh1a4, Aldhpb |
Location: |
Chr.19:20692953-20727556 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh1a7 | NM_011921.2 | NP_036051.1 | ||
| BC046315.1 | ||||
| BC139412.1 | ||||
| BC139413.1 | ||||
| U96401.1 | AAB64411.1 |
More Information
Set Membership: |
|

Genomic Map