Assay Details
Target Gene Details
Entrez Gene ID: | 50796 |
Gene Name: | doublesex and mab-3 related transcription factor 1 |
Gene Aliases: |
- |
Location: |
Chr.19:25505618-25604328 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmrt1 | NM_015826.5 | 2 | 544 | NP_056641.2 |
| AB221653.1 | 2 | 396 | ||
| AF192561.1 | 2 | 87 | AAF05764.1 | |
| AF202778.1 | 2 | 531 | AAF12826.1 | |
| AL133300.1 | 2 | 547 | CAB62040.1 | |
| AY169785.1 | 2 | 547 | AAO41733.1 | |
| AY169786.1 | 2 | 547 | AAO41734.1 | |
| AY169787.1 | 2 | 547 | AAO41735.1 | |
| AY943925.1 | AAY20941.1 | |||
| BC125474.1 | 2 | 507 | ||
| DQ530630.1 | 2 | 543 |
More Information
Set Membership: |
|

Genomic Map