Assay Details
Target Gene Details
Entrez Gene ID: | 57431 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 4 |
Gene Aliases: |
2010301J22Rik, Hspf2, Mcg18 |
Location: |
Chr.19:6987911-6992696 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc4 | NM_020566.1 | NP_065591.1 | ||
| AF036875.1 | AAC40117.1 | |||
| AK008507.1 | BAB25707.1 | |||
| AK170008.1 | BAE41509.1 | |||
| BC049281.1 | AAH49281.1 |

Genomic Map