Assay Details
Target Gene Details
Entrez Gene ID: | 236149 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 26 |
Gene Aliases: |
- |
Location: |
Chr.19:7781980-7802667 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a26 | NM_146232.1 | NP_666344.1 | ||
| BC014805.1 | AAH14805.1 |
More Information
Set Membership: |
|

Genomic Map