Assay Details
Target Gene Details
Entrez Gene ID: | 104252 |
Gene Name: | CDC42 effector protein (Rho GTPase binding) 2 |
Gene Aliases: |
1110008C05Rik, Borg1, Cep2, D19Bwg1013e |
Location: |
Chr.19:5917556-5924816 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42ep2 | NM_026772.2 | NP_081048.1 | ||
| BB842420.1 |

Genomic Map