Assay Details
Target Gene Details
Entrez Gene ID: | 16452 |
Gene Name: | Janus kinase 2 |
Gene Aliases: |
Fd17 |
Location: |
Chr.19:29251776-29313080 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jak2 | NM_001048177.2 | 18 | 2839 | NP_001041642.1 |
| NM_008413.3 | 19 | 2922 | NP_032439.2 | |
| BC054807.1 | 18 | 2602 | AAH54807.1 | |
| BC059834.1 | 18 | 2602 | AAH59834.1 | |
| L16956.1 | 18 | 2591 | AAB41327.1 |

Genomic Map