Assay Details
Target Gene Details
Entrez Gene ID: | 68423 |
Gene Name: | ankyrin repeat domain 13 family, member D |
Gene Aliases: |
0710001P18Rik, AI430801 |
Location: |
Chr.19:4270180-4283482 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd13d | NM_026720.2 | NP_080996.2 | ||
| AK089186.1 | ||||
| BC058982.1 | AAH58982.1 |

Genomic Map