Assay Details
Target Gene Details
Entrez Gene ID: | 52004 |
Gene Name: | CDK2-associated protein 2 |
Gene Aliases: |
5830466O21Rik, D19Ertd144e, Doc-1r |
Location: |
Chr.19:4096843-4099019 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk2ap2 | NM_026373.4 | NP_080649.1 | ||
| AK003686.2 | BAB22939.1 | |||
| AK012121.1 | BAB28046.1 | |||
| AK018035.1 | BAB31046.1 | |||
| BC025656.1 | AAH25656.1 |
More Information
Set Membership: |
|

Genomic Map