Assay Details
Target Gene Details
Entrez Gene ID: | 14283 |
Gene Name: | fos-like antigen 1 |
Gene Aliases: |
AW538199, Fra1, fra-1 |
Location: |
Chr.19:5447698-5455938 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fosl1 | NM_010235.2 | 2 | 416 | NP_034365.1 |
| AK144785.1 | 2 | 568 | BAE26065.1 | |
| AK144839.1 | 2 | 412 | BAE26093.1 | |
| AK145035.1 | 2 | 410 | BAE26198.1 | |
| BC052917.1 | 2 | 378 | AAH52917.1 | |
| U34245.1 | 2 | 359 | AAC52888.1 |
More Information
Set Membership: |
|

Genomic Map