Assay Details
Target Gene Details
Entrez Gene ID: | 240514 |
Gene Name: | coiled-coil domain containing 85B |
Gene Aliases: |
AI842788, BB115872 |
Location: |
Chr.19:5453163-5457563 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc85b | NM_001243307.1 | 2 | 2861 | NP_001230236.1 |
| NM_198616.4 | 1 | 3016 | NP_941018.1 | |
| AK140354.1 | 1 | 3348 |
Target Gene Details
Entrez Gene ID: | 14283 |
Gene Name: | fos-like antigen 1 |
Gene Aliases: |
AW538199, Fra1, fra-1 |
Location: |
Chr.19:5447698-5455938 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fosl1 | NM_010235.2 | NP_034365.1 | ||
| AK144785.1 | BAE26065.1 | |||
| AK144839.1 | BAE26093.1 | |||
| AK145035.1 | BAE26198.1 | |||
| BC052917.1 | AAH52917.1 | |||
| U34245.1 | AAC52888.1 |
More Information
Set Membership: |
|

Genomic Map