Assay Details
Target Gene Details
Entrez Gene ID: | 14869 |
Gene Name: | glutathione S-transferase, pi 2 |
Gene Aliases: |
GSTpiA, Gst-3, Gst3 |
Location: |
Chr.19:4040288-4042221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstp2 | NM_181796.2 | NP_861461.1 | ||
| BC064781.1 | AAH64781.1 | |||
| BC094623.1 | AAH94623.1 | |||
| GQ181117.1 |
More Information
Set Membership: |
|

Genomic Map