Assay Details
Target Gene Details
Entrez Gene ID: | 16452 |
Gene Name: | Janus kinase 2 |
Gene Aliases: |
Fd17 |
Location: |
Chr.19:29251776-29313080 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jak2 | NM_001048177.2 | 2 | 513 | NP_001041642.1 |
| NM_008413.3 | 3 | 596 | NP_032439.2 | |
| AK087255.1 | 2 | 478 | ||
| BC054807.1 | 2 | 276 | AAH54807.1 | |
| BC059834.1 | 2 | 276 | AAH59834.1 | |
| BE981299.1 | 3 | 401 | ||
| L16956.1 | 2 | 265 | AAB41327.1 |

Genomic Map