Assay Details
Target Gene Details
Entrez Gene ID: | 16452 |
Gene Name: | Janus kinase 2 |
Gene Aliases: |
Fd17 |
Location: |
Chr.19:29251776-29313080 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jak2 | NM_001048177.2 | 5 | 845 | NP_001041642.1 |
| NM_008413.3 | 6 | 928 | NP_032439.2 | |
| AK087255.1 | 5 | 811 | ||
| BC054807.1 | 5 | 608 | AAH54807.1 | |
| BC059834.1 | 5 | 608 | AAH59834.1 | |
| L16956.1 | 5 | 597 | AAB41327.1 |

Genomic Map