Assay Details
Target Gene Details
Entrez Gene ID: | 107351 |
Gene Name: | KN motif and ankyrin repeat domains 1 |
Gene Aliases: |
A930031B09Rik, AU015049, AW121052, Ankrd15, D330024H06Rik, mKIAA0172 |
Location: |
Chr.19:25236732-25434498 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kank1 | NM_181404.5 | NP_852069.4 | ||
| AK084638.1 | BAC39237.1 | |||
| AK122214.1 | BAC65496.2 | |||
| BC050250.1 | AAH50250.1 | |||
| BC058673.1 | AAH58673.1 | |||
| BC079563.1 | AAH79563.1 |
More Information
Set Membership: |
|

Genomic Map