Assay Details
Target Gene Details
Entrez Gene ID: | 12631 |
Gene Name: | cofilin 1, non-muscle |
Gene Aliases: |
AA959946, Cof, n-cofilin |
Location: |
Chr.19:5490455-5494031 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfl1 | NM_007687.5 | NP_031713.1 | ||
| AK013943.1 | BAB29074.1 | |||
| AK020471.1 | BAB32114.1 | |||
| AK088633.1 | BAC40467.1 | |||
| AK088793.1 | BAC40575.1 | |||
| AK167795.1 | BAE39825.1 | |||
| BC046225.1 | AAH46225.1 | |||
| BC058726.1 | AAH58726.1 | |||
| BC094357.1 | AAH94357.1 | |||
| CX218255.1 | ||||
| D00472.1 | BAA00364.1 |
More Information
Set Membership: |
|

Genomic Map