Assay Details
Target Gene Details
Entrez Gene ID: | 107351 |
Gene Name: | KN motif and ankyrin repeat domains 1 |
Gene Aliases: |
A930031B09Rik, AU015049, AW121052, Ankrd15, D330024H06Rik, mKIAA0172 |
Location: |
Chr.19:25236732-25434498 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kank1 | NM_181404.5 | NP_852069.4 | ||
| AK020911.1 | ||||
| AK041011.1 | BAC30778.1 |
More Information
Set Membership: |
|

Genomic Map