Assay Details
Target Gene Details
Entrez Gene ID: | 76002 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 18 |
Gene Aliases: |
5033428B15Rik |
Location: |
Chr.19:10997024-11018026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a18 | NM_001251849.1 | NP_001238778.1 | ||
| AK017199.1 |
More Information
Set Membership: |
|

Genomic Map