Assay Details
Target Gene Details
Entrez Gene ID: | 226162 |
Gene Name: | deleted in primary ciliary dyskinesia |
Gene Aliases: |
5330431N19Rik, Ndac |
Location: |
Chr.19:45560590-45578287 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpcd | NM_172639.2 | 2 | 129 | NP_766227.1 |
| AK077347.1 | 2 | 130 | BAC36762.1 | |
| BC027559.1 | 2 | 141 | AAH27559.1 |

Genomic Map