Assay Details
Target Gene Details
Entrez Gene ID: | 56642 |
Gene Name: | ankyrin repeat domain 2 (stretch responsive muscle) |
Gene Aliases: |
Arpp, mArpp |
Location: |
Chr.19:42035656-42045111 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd2 | NM_020033.1 | NP_064417.1 | ||
| AB076256.1 | BAB88557.1 | |||
| AJ011118.1 | CAB46646.1 | |||
| AJ245514.1 | CAB99431.1 | |||
| AK132585.1 | BAE21246.1 | |||
| BC138760.1 | ||||
| BC138761.1 |

Genomic Map