Assay Details
Target Gene Details
Entrez Gene ID: | 67116 |
Gene Name: | CUE domain containing 2 |
Gene Aliases: |
3010002G01Rik |
Location: |
Chr.19:46329812-46338660 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cuedc2 | NM_001164290.1 | NP_001157762.1 | ||
| NM_001164291.1 | NP_001157763.1 | |||
| NM_001164292.1 | NP_001157764.1 | |||
| NM_001164293.1 | NP_001157765.1 | |||
| NM_001164294.1 | NP_001157766.1 | |||
| NM_001164295.1 | NP_001157767.1 | |||
| NM_024192.2 | NP_077154.2 | |||
| AB232359.1 | BAE19943.1 | |||
| AK003457.1 | ||||
| AK008842.1 | BAB25923.3 | |||
| AK013884.1 | BAB29035.1 | |||
| AK186231.1 | ||||
| AK207375.1 | ||||
| BC003442.1 | AAH03442.1 | |||
| BY279028.1 | ||||
| BY312150.1 | ||||
| CK625575.1 | ||||
| CX218143.1 | ||||
| CX223880.1 |

Genomic Map