Assay Details
Target Gene Details
Entrez Gene ID: | 319594 |
Gene Name: | hypoxia-inducible factor 1, alpha subunit inhibitor |
Gene Aliases: |
2310046M24Rik, A830014H24Rik, FIH, FIH1 |
Location: |
Chr.19:44562854-44576274 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hif1an | NM_176958.3 | 1 | 137 | NP_795932.2 |
| AK043636.1 | 1 | 123 | BAC31602.1 | |
| AK084960.1 | 1 | 132 | ||
| AK141867.1 | 1 | 123 | ||
| AK154774.1 | 1 | 137 | BAE32822.1 | |
| BC130013.1 | 1 | 141 | ||
| BY122345.1 | 1 | 141 | ||
| BY713373.1 | 1 | 123 |

Genomic Map