Assay Details
Target Gene Details
Entrez Gene ID: | 19697 |
Gene Name: | v-rel reticuloendotheliosis viral oncogene homolog A (avian) |
Gene Aliases: |
p65 |
Location: |
Chr.19:5637440-5648130 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rela | NM_009045.4 | NP_033071.1 | ||
| AK089489.1 | BAC40904.1 | |||
| AK144565.1 | ||||
| AK154630.1 | BAE32727.1 | |||
| AK155321.1 | ||||
| AK155889.1 | BAE33485.1 | |||
| AK157076.1 | BAE33954.1 | |||
| AK158072.1 | BAE34348.1 | |||
| BC003818.1 | AAH03818.1 | |||
| BC094053.1 | AAH94053.1 | |||
| DQ020177.1 | AAY44745.1 | |||
| M61909.1 | AAA39811.1 |
More Information
Set Membership: |
|

Genomic Map