Assay Details
Target Gene Details
Entrez Gene ID: | 207151 |
Gene Name: | solute carrier family 22 (organic anion transporter), member 19 |
Gene Aliases: |
D630043A20Rik, OAT-5, Oat5, Slc22a9 |
Location: |
Chr.19:7672654-7711310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a19 | NM_144785.2 | NP_659034.1 | ||
| AF529221.1 | AAQ09529.1 | |||
| AK052752.1 | ||||
| BC021384.1 | AAH21384.1 |
More Information
Set Membership: |
|

Genomic Map