Assay Details
Target Gene Details
Entrez Gene ID: | 20867 |
Gene Name: | stress-induced phosphoprotein 1 |
Gene Aliases: |
Hop, Sti1, p60 |
Location: |
Chr.19:7020696-7040026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stip1 | NM_016737.2 | NP_058017.1 | ||
| AK075988.1 | BAC36100.1 | |||
| AK088494.1 | BAC40389.1 | |||
| AK149493.1 | BAE28916.1 | |||
| AK161645.1 | BAE36509.1 | |||
| AK167273.1 | BAE39385.1 | |||
| AK168181.1 | BAE40141.1 | |||
| AK178711.1 | ||||
| AK181085.1 | ||||
| BC003794.1 | AAH03794.1 | |||
| U27830.1 | AAC53267.1 |
More Information
Set Membership: |
|

Genomic Map