Assay Details
Target Gene Details
Entrez Gene ID: | 109225 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 7 |
Gene Aliases: |
9130422I10Rik, A430103C15Rik, CD20l4, CFFMA |
Location: |
Chr.19:11321039-11336146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a7 | NM_001025610.4 | NP_001020781.1 | ||
| NM_001276398.1 | NP_001263327.1 | |||
| NM_027836.6 | NP_082112.3 | |||
| AF237917.2 | AAK37607.2 | |||
| AK018686.1 | BAB31344.1 | |||
| AK046559.1 | ||||
| AK079888.1 | BAC37776.1 | |||
| AK085297.1 | BAC39413.1 | |||
| AK149890.1 | BAE29148.1 | |||
| AK151261.1 | BAE30249.1 | |||
| AK152098.1 | BAE30945.1 | |||
| BC024402.1 | AAH24402.1 |
More Information
Set Membership: |
|

Genomic Map