Assay Details
Target Gene Details
Entrez Gene ID: | 70166 |
Gene Name: | lipase, family member N |
Gene Aliases: |
2210418G03Rik, Lipl4 |
Location: |
Chr.19:34067058-34085946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lipn | NM_027340.2 | NP_081616.1 | ||
| AK008974.1 | BAB26000.1 | |||
| AK154333.1 | BAE32519.1 |
More Information
Set Membership: |
|

Genomic Map