Assay Details
Target Gene Details
Entrez Gene ID: | 23942 |
Gene Name: | metastasis-associated gene family, member 2 |
Gene Aliases: |
AW550797, Mata1l1, Mta1l1, mmta2 |
Location: |
Chr.19:8941920-8952300 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mta2 | NM_011842.3 | NP_035972.3 | ||
| AF159259.1 | AAD51281.1 | |||
| AK077548.1 | ||||
| AK088158.1 | BAC40180.1 | |||
| AK147099.1 | BAE27675.1 | |||
| AK149834.1 | BAE29113.1 | |||
| AK157717.1 | BAE34165.1 | |||
| AK159979.1 | BAE35530.1 | |||
| AK180159.1 | ||||
| AK183116.1 | ||||
| AK185749.1 | ||||
| AK206505.1 | ||||
| BC079847.1 | AAH79847.1 |
More Information
Set Membership: |
|

Genomic Map