Assay Details
Target Gene Details
Entrez Gene ID: | 226115 |
Gene Name: | oligodendrocytic myelin paranodal and inner loop protein |
Gene Aliases: |
Tmem10, Tmp10 |
Location: |
Chr.19:41063420-41077113 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Opalin | NM_153520.1 | NP_705740.1 | ||
| BC060980.1 | AAH60980.1 |
More Information
Set Membership: |
|

Genomic Map