Assay Details
Target Gene Details
Entrez Gene ID: | 28000 |
Gene Name: | PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae) |
Gene Aliases: |
AA617263, AL024362, D19Wsu55e, NMP200, PSO4, Prp19, Snev |
Location: |
Chr.19:10895231-10909559 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf19 | NM_001253843.1 | NP_001240772.1 | ||
| NM_134129.4 | NP_598890.1 | |||
| AB222602.1 | BAE16340.1 | |||
| AF251503.1 | AAK49039.1 | |||
| AK047693.1 | BAC33127.1 | |||
| AK088769.1 | BAC40560.1 | |||
| AK164681.1 | BAE37873.1 | |||
| AK182400.1 | ||||
| AK182674.1 | ||||
| AK184270.1 | ||||
| AK185414.1 | ||||
| AK185550.1 | ||||
| AK185866.1 | ||||
| AK188848.1 | ||||
| BC004070.1 | AAH04070.1 |
More Information
Set Membership: |
|

Genomic Map