Assay Details
Target Gene Details
Entrez Gene ID: | 225998 |
Gene Name: | RAR-related orphan receptor beta |
Gene Aliases: |
Nr1f2, RZR-beta, RZRB, Rorbeta |
Location: |
Chr.19:18930609-19111196 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rorb | NM_001043354.2 | NP_001036819.1 | ||
| NM_001289921.1 | NP_001276850.1 | |||
| NM_146095.4 | NP_666207.3 | |||
| AB258405.1 | ||||
| AK134790.1 | ||||
| BC024842.1 | ||||
| BC058269.1 | AAH58269.1 | |||
| DQ779923.1 | ||||
| DQ779924.1 |
More Information
Set Membership: |
|

Genomic Map