Assay Details
Target Gene Details
Entrez Gene ID: | 56389 |
Gene Name: | syntaxin 5A |
Gene Aliases: |
0610031F24Rik, D19Ertd627e, Stx5 |
Location: |
Chr.19:8741424-8755642 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx5a | NM_001167799.1 | NP_001161271.1 | ||
| NM_019829.4 | NP_062803.4 | |||
| AF232709.1 | AAF36981.1 | |||
| AK002722.1 | ||||
| AK166087.1 | BAE38563.1 | |||
| AK181717.1 | 1 | 129 | ||
| AK190896.1 | ||||
| AK201004.1 | ||||
| AK212010.1 | 1 | 114 | ||
| BC004849.1 | AAH04849.1 | |||
| BC021883.1 | AAH21883.1 | |||
| CN539678.1 |
More Information
Set Membership: |
|

Genomic Map