Assay Details
Target Gene Details
Entrez Gene ID: | 73166 |
Gene Name: | transmembrane 7 superfamily member 2 |
Gene Aliases: |
3110041O18Rik, ANG1, C14SR, Dhcr14 |
Location: |
Chr.19:6062821-6067850 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm7sf2 | NM_028454.2 | NP_082730.2 | ||
| AK014166.1 | BAB29187.1 | |||
| AK037325.1 | ||||
| AK039028.1 | ||||
| AK040016.1 | BAE20564.1 | |||
| AK043687.1 | 1 | 35 | ||
| AK134512.1 | ||||
| AK155475.1 | BAE33285.1 | |||
| BC014769.1 | AAH14769.1 |
Target Gene Details
Entrez Gene ID: | 68505 |
Gene Name: | vacuolar protein sorting 51 homolog (S. cerevisiae) |
Gene Aliases: |
1110014N23Rik, 3110057M17Rik, AI837850, Ffr |
Location: |
Chr.19:6067842-6077187 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps51 | NM_001081041.1 | NP_001074510.1 |
More Information
Set Membership: |
|

Genomic Map